ContactUsLinkComponent
Rabbit IgG polyclonal antibody for Cytoskeleton-associated protein 4(Ckap4) detection. Tested with WB in Mouse. 1 * 100 µG
Catalog # BSBTA02154-1
Supplier:  Boster Bio
undefined
Rabbit IgG polyclonal antibody for Cytoskeleton-associated protein 4(Ckap4) detection. Tested with WB in Mouse. 1 * 100 µG
Catalog # BSBTA02154-1
Supplier:  Boster Bio