ContactUsLinkComponent
Rabbit IgG polyclonal antibody for A disintegrin and metalloproteinase with thrombospondin motifs 1(ADAMTS1) detection. Tested with WB in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA03009-1
Supplier:  Boster Bio
undefined
Rabbit IgG polyclonal antibody for A disintegrin and metalloproteinase with thrombospondin motifs 1(ADAMTS1) detection. Tested with WB in Human Mouse Rat. 1 * 100 µG
Catalog # BSBTA03009-1
Supplier:  Boster Bio