ContactUsLinkComponent
For quantitative detection of human ULBP2 in cell culture supernates, cell lysates, tissue homogenates, serum and plasma (heparin, E DTA). 1 * 1 KIT
Catalog # BSBTEK1593
Supplier:  Boster Bio
undefined
For quantitative detection of human ULBP2 in cell culture supernates, cell lysates, tissue homogenates, serum and plasma (heparin, E DTA). 1 * 1 KIT
Catalog # BSBTEK1593
Supplier:  Boster Bio