ContactUsLinkComponent
Stackable container with reinforcing cross-bars. Capacity 530 L. Made of High-density polyethylene. For foods. Load capacity: 250 Kg. Outside dimensions 1200 x 800 x 960 mm. Inside dimensions 1100 x 735 x 650 mm. Color,,Grey. With rubber wheels Ø 125 1 * 1 items
Catalog # LAMA1428CR
Supplier:  LAMAPLAST
Stackable container with reinforcing cross-bars. Capacity 530 L. Made of High-density polyethylene. For foods. Load capacity: 250 Kg. Outside dimensions 1200 x 800 x 960 mm. Inside dimensions 1100 x 735 x 650 mm. Color,,Grey. With rubber wheels Ø 125 1 * 1 items
Catalog # LAMA1428CR
Supplier:  LAMAPLAST
Frequently Bought Together