ContactUsLinkComponent
a blue protein standard with 12 prestained proteins covering a wide range of molecular weights:10-240kda in tris-glycine buffer and 9-235kda in bis-tris (mops) buffer or bis-tris (mes) buffer. proteins are covalently coupled with a blue chromophore 1 * 100 items
Catalog # GENO786-418
Supplier:  G-Biosciences
a blue protein standard with 12 prestained proteins covering a wide range of molecular weights:10-240kda in tris-glycine buffer and 9-235kda in bis-tris (mops) buffer or bis-tris (mes) buffer. proteins are covalently coupled with a blue chromophore 1 * 100 items
Catalog # GENO786-418
Supplier:  G-Biosciences