ContactUsLinkComponent
g-trap ida agarose fast flow columns are ready to use prepacked affinity columns used for purification of proteins based on immobil ized metal ion affinity chromatography (imac). imac is based on interaction of cations of transition metals such as n 1 * 5 mL
Catalog # GENO786-1022
Supplier:  G-Biosciences
g-trap ida agarose fast flow columns are ready to use prepacked affinity columns used for purification of proteins based on immobil ized metal ion affinity chromatography (imac). imac is based on interaction of cations of transition metals such as n 1 * 5 mL
Catalog # GENO786-1022
Supplier:  G-Biosciences