ContactUsLinkComponent
Viewing window and interior lighting - 230 V option model, viewing window (triple insulating glass) in door, 370 x 370 mm, and 15 W interior 1 * 1 items
Catalog # BIND8012-1708
Supplier:  Binder
Viewing window and interior lighting - 230 V option model, viewing window (triple insulating glass) in door, 370 x 370 mm, and 15 W interior 1 * 1 items
Catalog # BIND8012-1708
Supplier:  Binder
Frequently Bought Together