ContactUsLinkComponent
Long Path Rectangular Cell Holder on G10 Base Holds rectangular cells 10mm to 100mm Fits all GENESYS 10/10S, BioMate 3/3S, Evolution 60/60S Spectrophotometers 1 * 1 items
Catalog # SPET840-284200
Long Path Rectangular Cell Holder on G10 Base Holds rectangular cells 10mm to 100mm Fits all GENESYS 10/10S, BioMate 3/3S, Evolution 60/60S Spectrophotometers 1 * 1 items
Catalog # SPET840-284200
Frequently Bought Together