ContactUsLinkComponent
The Thermo Scientific™ High-Select™ TiO2 Phosphopeptide Enrichment Kit enables efficient enrichment of phosphorylated peptides from complex and fractionated protein digests for analysis by mass spectrometry (MS). 1 * 24 AU
Catalog # PIERA32993
The Thermo Scientific™ High-Select™ TiO2 Phosphopeptide Enrichment Kit enables efficient enrichment of phosphorylated peptides from complex and fractionated protein digests for analysis by mass spectrometry (MS). 1 * 24 AU
Catalog # PIERA32993
Frequently Bought Together