ContactUsLinkComponent
Flask for collecting 1l, with cylindrical needle valve,st29/32,300x80mmmm, acc. to schlenk, null
Catalog # WITG0.778.303.V
Supplier:  witeg Labortechnik
Flask for collecting 1l, with cylindrical needle valve,st29/32,300x80mmmm, acc. to schlenk, null
Catalog # WITG0.778.303.V
Supplier:  witeg Labortechnik
Frequently Bought Together