ContactUsLinkComponent
Flask for collecting 50ml, with cylindrical needle valve,145x32mmmm, acc. to schlenk, null
Catalog # WITG0.778.203.V
Supplier:  witeg Labortechnik
Flask for collecting 50ml, with cylindrical needle valve,145x32mmmm, acc. to schlenk, null
Catalog # WITG0.778.203.V
Supplier:  witeg Labortechnik
Frequently Bought Together