ContactUsLinkComponent
Flask for collecting nitrogen 1l, 300x80mm, st29/32,acc. to schlenk, null
Catalog # WITG0.778.103
Supplier:  witeg Labortechnik
Flask for collecting nitrogen 1l, 300x80mm, st29/32,acc. to schlenk, null
Catalog # WITG0.778.103
Supplier:  witeg Labortechnik
Frequently Bought Together