ContactUsLinkComponent
Tubing threaded 75x130x2.2mm gl60 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
Catalog # WITG0.183.060
Supplier:  witeg Labortechnik
Tubing threaded 75x130x2.2mm gl60 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
Catalog # WITG0.183.060
Supplier:  witeg Labortechnik
Frequently Bought Together