ContactUsLinkComponent
Tubing threaded 26x110x2.0mm gl25 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
  WITG0.183.025
Tubing threaded 26x110x2.0mm gl25 ,with restriction,acc. to din 12216, with iso screw thread,made of borosilicate glass 1 * 1 items
  WITG0.183.025
Frequently Bought Together