ContactUsLinkComponent
Tubing threaded 22x110x1.8mm svl22 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl 1 * 1 items
Catalog # WITG0.182.322
Supplier:  witeg Labortechnik
Tubing threaded 22x110x1.8mm svl22 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl 1 * 1 items
Catalog # WITG0.182.322
Supplier:  witeg Labortechnik
Frequently Bought Together