ContactUsLinkComponent
Tubing threaded 8x110x1.5mm rd10 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl null
Catalog # WITG0.182.210
Supplier:  witeg Labortechnik
Tubing threaded 8x110x1.5mm rd10 with screw threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, available with screw thread gl, rd or svl null
Catalog # WITG0.182.210
Supplier:  witeg Labortechnik
Frequently Bought Together