ContactUsLinkComponent
Tubing threaded 95x130x3.5mm gl100 with screw,threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.100
Supplier:  witeg Labortechnik
Tubing threaded 95x130x3.5mm gl100 with screw,threaded straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.100
Supplier:  witeg Labortechnik
Frequently Bought Together