ContactUsLinkComponent
Tubing threaded 65x130x3.2mm gl70 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.070
Supplier:  witeg Labortechnik
Tubing threaded 65x130x3.2mm gl70 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.070
Supplier:  witeg Labortechnik
Frequently Bought Together