ContactUsLinkComponent
Tubing threaded 56x130x2.5mm gl60 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.060
Supplier:  witeg Labortechnik
Tubing threaded 56x130x2.5mm gl60 with screw threaded, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass , available with screw thread gl, rd or svl null
Catalog # WITG0.182.060
Supplier:  witeg Labortechnik
Frequently Bought Together