ContactUsLinkComponent
Tubing threaded 28x150x2.0mm gl32 with screw, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, availab le with screw thread gl, rd or svl null
Catalog # WITG0.182.032
Supplier:  witeg Labortechnik
Tubing threaded 28x150x2.0mm gl32 with screw, straight,acc. to din 12216, with iso screw thread,made of borosilicate glass, availab le with screw thread gl, rd or svl null
Catalog # WITG0.182.032
Supplier:  witeg Labortechnik
Frequently Bought Together