ContactUsLinkComponent
SIK3 antibody was raised against an 18 amino acid synthetic peptide nearthe carboxy terminus of human SIK3. 1 * 100 µG
Catalog # NOVUNBP2-47277
Supplier:  Novus Biologicals
SIK3 antibody was raised against an 18 amino acid synthetic peptide nearthe carboxy terminus of human SIK3. 1 * 100 µG
Catalog # NOVUNBP2-47277
Supplier:  Novus Biologicals
Frequently Bought Together