ContactUsLinkComponent
Round contact Petri plate PS na3 ve nts H14.5 Ø65 & grip lid, 20/bag easy to open, double outer bag with traceability, STERILE R (Number of the sub-conditionning unit : 20 petri plates Quantity per carton : 720) 1 * 720 items
  GOSSBP63-08
 :  Corning
Round contact Petri plate PS na3 ve nts H14.5 Ø65 & grip lid, 20/bag easy to open, double outer bag with traceability, STERILE R (Number of the sub-conditionning unit : 20 petri plates Quantity per carton : 720) 1 * 720 items
  GOSSBP63-08
 :  Corning
Frequently Bought Together