EC HPLC column EC 250/4.6 NUCLEOSIL 1000-7 NH2 length: 250 mm, ID: 4.6 mm pack of 1 1 * 1 items
MANA720363.46
EC HPLC column EC 250/4.6 NUCLEOSIL 1000-7 NH2 length: 250 mm, ID: 4.6 mm pack of 1 1 * 1 items
MANA720363.46
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection