ContactUsLinkComponent
550 ph and temperature meter portable, supplied with epoxy bodied electrode (924001), atc probe and buffers in a handy carry case 1 * 1 items
Catalog # JENW10088-53
Supplier:  Cole-Parmer
550 ph and temperature meter portable, supplied with epoxy bodied electrode (924001), atc probe and buffers in a handy carry case 1 * 1 items
Catalog # JENW10088-53
Supplier:  Cole-Parmer
Frequently Bought Together