ContactUsLinkComponent
Biological Indicator Spore Strips for Monitoring Steam Processes, Geobacillus stearothermophilus Cell Line 7953, 7 mm x 38 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 105 population level, ISO 11138-1, 11138-3 and USP com 1 * 100 items
Catalog # NAMSBS-105
Supplier:  CROSSTEX
Biological Indicator Spore Strips for Monitoring Steam Processes, Geobacillus stearothermophilus Cell Line 7953, 7 mm x 38 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 105 population level, ISO 11138-1, 11138-3 and USP com 1 * 100 items
Catalog # NAMSBS-105
Supplier:  CROSSTEX