ContactUsLinkComponent
Thermometer precision, measuring range: -80 + 20 ° C in 1 ° C density-wide range,IP 2 C Low Cloud and Pour, capillary: yellow coate d round Display liquid: toluene red immersion depth: 76mm, total length: 225 mm with DAkkS calibration ASTM 6 C - ACCU 1 * 1 items
Catalog # SCNL1202006/04/3P
Thermometer precision, measuring range: -80 + 20 ° C in 1 ° C density-wide range,IP 2 C Low Cloud and Pour, capillary: yellow coate d round Display liquid: toluene red immersion depth: 76mm, total length: 225 mm with DAkkS calibration ASTM 6 C - ACCU 1 * 1 items
Catalog # SCNL1202006/04/3P
Frequently Bought Together