ContactUsLinkComponent
External humidity and temperature probe plug-in w/o cable will connect to testo 176 H1/H2/P1, testo Saveris H4D, testo Saveris 2-H2 diameter 12mm. 1 * 1 items
Catalog # TEST0572.2151
Supplier:  Testo
External humidity and temperature probe plug-in w/o cable will connect to testo 176 H1/H2/P1, testo Saveris H4D, testo Saveris 2-H2 diameter 12mm. 1 * 1 items
Catalog # TEST0572.2151
Supplier:  Testo
Frequently Bought Together