ContactUsLinkComponent
Drop-n-Stain CF488A donkey anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, 1 * 5 mL
Catalog # BTIU20952
Supplier:  Biotium
Drop-n-Stain CF488A donkey anti-mouse IgG (H+L) is provided in a convenient dropper bottle for easy dispensing without pipetting. To minimize cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, 1 * 5 mL
Catalog # BTIU20952
Supplier:  Biotium
Frequently Bought Together