PAH-Mix 9, each µg/mL in Acetonitrile, 10 mL Dibenzo[a,h]anthracen 0,5 µg/ml Benzo[a]anthracen 1,0 µg/ml 6-Methylchrysen 1,0 µg/ml Benzo[e]pyren 1,0 µg/ml Benzo[j]fluoranthen 1,0 µg/ml Benzo[k]fluoranthen 1,0 µg/ml Benzo[a]pyren 1,0 µg/ml Benzo[b]flu 1 * 10 ml
Katalogové číslo PROOSL24210
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together