ContactUsLinkComponent
Base model without HEPA, without Accessories, for dry recovery only Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity:23L - Dry recovery 28L – Wet Dimensions: H81cm×W48cm×L61cm Noise level: 70 dB (A) 1 * 1 items
  TIGV110262
 :  TIGER-VAC
Base model without HEPA, without Accessories, for dry recovery only Air flow: 187 m3/h Vacuum pressure: 4572 mmH2O Recovery Capacity:23L - Dry recovery 28L – Wet Dimensions: H81cm×W48cm×L61cm Noise level: 70 dB (A) 1 * 1 items
  TIGV110262
 :  TIGER-VAC
Frequently Bought Together