ContactUsLinkComponent
Made of soft, breathable, three layer material, Foliodrape® Arm covers have elastic knit cuffs and the double layer is firmly sewn on, with sealed sleeve seams and an elastic rim giving a secure fit on the upper arm. They are impermeable to moisture 1 * 1 PAC
  HART258330
Made of soft, breathable, three layer material, Foliodrape® Arm covers have elastic knit cuffs and the double layer is firmly sewn on, with sealed sleeve seams and an elastic rim giving a secure fit on the upper arm. They are impermeable to moisture 1 * 1 PAC
  HART258330
Frequently Bought Together