ContactUsLinkComponent
24 components Al 300µg/l Sb 30µg/l As 15µg/l Ba 30µg/l Be 3µg/l B 300µg/l Cd 2.4µg/l Cr 30µg/l Fe 300µg/l Co 1.8µg/l Cu 30µg/l Pb 30µg/l Mn 30µg/l Mo 30µg/l Ni 30µg/l Se 15µg/l Te 30µg/l Tl 3µg/l 1 * 500 mL
Catalog # CMPLF424.D0035N.L5
24 components Al 300µg/l Sb 30µg/l As 15µg/l Ba 30µg/l Be 3µg/l B 300µg/l Cd 2.4µg/l Cr 30µg/l Fe 300µg/l Co 1.8µg/l Cu 30µg/l Pb 30µg/l Mn 30µg/l Mo 30µg/l Ni 30µg/l Se 15µg/l Te 30µg/l Tl 3µg/l 1 * 500 mL
Catalog # CMPLF424.D0035N.L5
Frequently Bought Together