ContactUsLinkComponent
24 components Al 100µg/l Sb 10µg/l As 5µg/l Ba 10µg/l Be 1µg/l B 100µg/l Cd 0.8µg/l Cr 10µg/l Fe 100µg/l Co 0.6µg/l Cu 10µg/l Pb 10µg/l Mn 10µg/l Mo 10µg/l Ni 10µg/l Se 5µg/l Te 10µg/l Tl 1µg/l 1 * 100 mL
Catalog # CMPLF424.D0015N.L1
24 components Al 100µg/l Sb 10µg/l As 5µg/l Ba 10µg/l Be 1µg/l B 100µg/l Cd 0.8µg/l Cr 10µg/l Fe 100µg/l Co 0.6µg/l Cu 10µg/l Pb 10µg/l Mn 10µg/l Mo 10µg/l Ni 10µg/l Se 5µg/l Te 10µg/l Tl 1µg/l 1 * 100 mL
Catalog # CMPLF424.D0015N.L1
Frequently Bought Together