Fluorescein hexylamine can be coupled to activated carboxy groups or sulfonyl chlorides. This product could be used as a direct rep lacement to fluorescein cadaverine (cat#: 92000) 1 * 25 mg
Catalog # 91058.
Supplier: Biotium
Frequently Bought Together