ContactUsLinkComponent
CF™583 maleimide is a superior deep red fluorescent dye with a maleimide reactive group, which can be used to label a target molecul e with a thiol group. CF™583 is spectrally identical to Cy®3.5 but yields much brighter protein conjugates. 1 * 1 µmol
Catalog # BTIU96017
Supplier:  Biotium
CF™583 maleimide is a superior deep red fluorescent dye with a maleimide reactive group, which can be used to label a target molecul e with a thiol group. CF™583 is spectrally identical to Cy®3.5 but yields much brighter protein conjugates. 1 * 1 µmol
Catalog # BTIU96017
Supplier:  Biotium
Frequently Bought Together