ContactUsLinkComponent
C9 is a plasma protein synthesized in the liver and monocytes consisting of a single polypeptide chain. C9 is a part of the membrane attack complex (MAC), an important component of the immune system. 1 * 100 µG
  BWRLBS61143
C9 is a plasma protein synthesized in the liver and monocytes consisting of a single polypeptide chain. C9 is a part of the membrane attack complex (MAC), an important component of the immune system. 1 * 100 µG
  BWRLBS61143
Frequently Bought Together