ContactUsLinkComponent
The mucins are a family of highly glycosylated, secreted proteins with a basic structure consisting of a variable number of tandem r epeats (VNTRs) encoded by 60 base pairs (Mucin 1), 69 base pairs (Mucin 2) and 51 base pairs (Mucin 3). 1 * 100 µG
  BWRLBS60935
The mucins are a family of highly glycosylated, secreted proteins with a basic structure consisting of a variable number of tandem r epeats (VNTRs) encoded by 60 base pairs (Mucin 1), 69 base pairs (Mucin 2) and 51 base pairs (Mucin 3). 1 * 100 µG
  BWRLBS60935
Frequently Bought Together