ContactUsLinkComponent
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of c hromosome condensation 1 (RCC1 MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MI 1 * 100 µG
Katalogové číslo BWRLBS8706
Dodavatel:  Bioworld Technology
The METTL11A gene encodes an N-terminal methyltransferase for the RAN (MIM 601179) guanine nucleotide exchange factor regulator of c hromosome condensation 1 (RCC1 MIM 179710). METTL11A enzyme alpha-N-methylates other protein targets such as SET (MI 1 * 100 µG
Katalogové číslo BWRLBS8706
Dodavatel:  Bioworld Technology
Frequently Bought Together