ContactUsLinkComponent
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membr ane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. 1 * 100 µG
  BWRLBS8701
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membr ane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. 1 * 100 µG
  BWRLBS8701
Frequently Bought Together