NKp46 (lymphocyte antigen 94, LY94) is a natural cytotoxicity receptor that belongs to the immunoglobulin superfamily and is express ed by all resting or activated NK cells, but not on T cells or B cells. 1 * 100 µG
Catalog # BWRLBS8525
Supplier: Bioworld Technology
Frequently Bought Together