Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
IPA 10 l, schnell, effektiv und rückstandsfrei gebrauchsfertig farbstoff- und parfümfrei Wirksamkeit geprüft nach Europäischen Normen (EN) 1 * 10 L
Katalogové číslo SCUE70000030
Supplier:
SCHUELKE MAYR
Certificates
Expand image
IPA 10 l, schnell, effektiv und rückstandsfrei gebrauchsfertig farbstoff- und parfümfrei Wirksamkeit geprüft nach Europäischen Normen (EN) 1 * 10 L
Katalogové číslo SCUE70000030
Supplier:
SCHUELKE MAYR
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Tetrahydrofuran, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Acetone, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Formaldehyde ≥37% in aqueous solution stabilised, GR ACS, Reag. Ph. Eur. analytical reagent, Sigma-Aldrich®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Detergent, powder for automated cleaning of glassware, neodisher® LaboClean A8
Supplier: Dr. Weigert
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals