ContactUsLinkComponent
For quantitative detection of rat CD56 in cell culture supernates, serum and plasma(heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1562
Supplier:  Boster Bio
undefined
For quantitative detection of rat CD56 in cell culture supernates, serum and plasma(heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1562
Supplier:  Boster Bio