ContactUsLinkComponent
For quantitative detection of rat ALCAM in cell culture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1572
Supplier:  Boster Bio
undefined
For quantitative detection of rat ALCAM in cell culture supernates, serum and plasma (heparin, EDTA). 1 * 1 KIT
Catalog # BSBTEK1572
Supplier:  Boster Bio