ContactUsLinkComponent
Mycoplasma pneumoniae IgM ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgM antibodies to Mycopla sma pneumoniae. 1 * 1 KIT
Catalog # ABNOKA4850
Supplier:  Abnova
Mycoplasma pneumoniae IgM ELISA Kit is an enzyme-linked immunosorbent assay for the detection of IgM antibodies to Mycopla sma pneumoniae. 1 * 1 KIT
Catalog # ABNOKA4850
Supplier:  Abnova
Frequently Bought Together