VIAL SCREW 2ML AMBR WRTON LP 1 * 100 items
Catalog # HEWL5190-4034
Supplier: Agilent
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Screw caps for screw neck vials, N 9
Supplier: MACHEREY-NAGEL
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
BD Discardit™ II Syringes
Supplier: BD Medical
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
ESD safety shoes, lace-up, uvex 1 ladies, 8562
Supplier: UVEX
Petroleum spirit, 180…280 °C ∼, pure
Supplier: Thermo Fisher Scientific