Tris solution is an abbreviation of the organic compound known as tris(hydroxymethyl)aminomethane. Tris is extensively used in biochemistry and molecular biology. In biochemistry, Tris is widely used as a component of buffer solutions, such as in TAE 1 * 1 L
Catalog # AVIVOORA00255
Supplier: Aviva Systems Biology
Frequently Bought Together