ContactUsLinkComponent
HEPA air filter - on fresh air supply for unit filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 µm) 1 * 1 items
Catalog # BIND8012-1051
Supplier:  Binder
HEPA air filter - on fresh air supply for unit filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 µm) 1 * 1 items
Catalog # BIND8012-1051
Supplier:  Binder
Frequently Bought Together