Sodium molybdate anhydrous, powder, _100 mesh particle size, 99.9% trace metals basis grade anhydrous assay 99.9% trace metals basis form powder particle size _100 mesh mp 687 C(lit.) density 3.78 g/mL at 25 C(lit.) 1 * 5 g
Catalog # SIAL737860-5G
Supplier: Merck
Frequently Bought Together