ContactUsLinkComponent
This product is prepared by labeling high quality goat anti-mouse IgG isotype-specific secondary antibody conjugates with biotin. Th e conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse Ig 1 * 250 µl
Catalog # 20473-250UL
Supplier:  Biotium
This product is prepared by labeling high quality goat anti-mouse IgG isotype-specific secondary antibody conjugates with biotin. Th e conjugates are prepared from affinity-purified antibodies that react with Fc portion of the heavy chain of mouse Ig 1 * 250 µl
Catalog # 20473-250UL
Supplier:  Biotium
Frequently Bought Together