ContactUsLinkComponent
ATEX CATEGORY 2 ZONES 1-2 (GAS) & ZONES 21-22 (DUST) MAXIMUM SURFACE TEMPERATURE : T6, 85°C AMBIANT TEMPERATURE : +40°C / -20°C Air Operated Vacuum Pressure: 4572 mm H2O Air flow: 187 m3/h Dry Recovery: 19 L Dimensions: H107cm×W71cm×L51cm Wei 1 * 1 items
Catalog # TIGV110400
Supplier:  TIGER-VAC
ATEX CATEGORY 2 ZONES 1-2 (GAS) & ZONES 21-22 (DUST) MAXIMUM SURFACE TEMPERATURE : T6, 85°C AMBIANT TEMPERATURE : +40°C / -20°C Air Operated Vacuum Pressure: 4572 mm H2O Air flow: 187 m3/h Dry Recovery: 19 L Dimensions: H107cm×W71cm×L51cm Wei 1 * 1 items
Catalog # TIGV110400
Supplier:  TIGER-VAC
Frequently Bought Together